Would you like to log in again?
RTW Adjustable-Height Worktables, Production Basics
Supplier: Production Basics
RTW
PB-3000 3102 PB-3105 PB-3128 PB-3023 3002 3011 3005 PB-3123-G PB-3001 3055 3030 3100 3115 3060 PB-3109 PB-3050 3105 PB-3123 3007 3057 3110 PB-3114 3001 3000 3107 3015 3052 3114 3111 PB-3003 PB-3110 3109 3010 3014 PB-3005 3050 3061 3103 3063
97008-310EA 566.33 USD InStock
97008-310 76245-968 76245-966 76246-236 76246-056 11005-152 76267-896 76239-042 11005-130 76240-230 87000-322 76239-040 11005-150 97008-308 87000-320 76267-892 76239-044 11005-138 11005-136 11005-134 87000-326 11005-132 87000-310 11005-142 11005-140 76240-226 87000-318 76268-318 11005-128 97008-312 87000-316 76240-228 11005-126 11005-148 87000-314 11005-124 11005-146 87000-312 11005-122 11005-144
RTW Adjustable-Height Worktables, Production Basics
Furniture Tables and Workbenches
Ergonomic tables are constructed of 12-gauge steel and industrial-grade laminate with an abrasion-resistant finish.
  • Optional ESD laminate has static dissipative range of 106 to 109
  • Height is adjustable in 5 cm (2") increments from 76.2 to 91.4 cm (30 to 36")
Standard models include blue frame with white laminate work surface or gray ESD laminate work surface.
Load capacity: 750 lbs.
Ordering information: Tables include frame, work surface, four 5×5 cm (2×2") leg adjusters, leveling feet, and parts and hardware packet for assembly. ESD laminate tables also include ground bolt and wire. To order uprights, accessories, or frames and work surfaces in different colors, contact your Avantor representative. Additional frame colors include light neutral, white, gray, and black; laminate colors include gray, almond, ESD beige, and ESD white.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty