Would you like to log in again?
Chlorine (Free and Total) Reagent Powder, DPD 10 ml, Lovibond®
Supplier: Tintometer
530120 530100
97009-912PK 36.32 USD InStock
97009-912 97009-910
Chlorine (Free and Total) Reagent Powder, DPD 10 ml, Lovibond®
Environmental Test Kits
For determination of free and total chlorine by the DPD method
  • Suitable for use with colorimeters and spectrophotometers
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty