Would you like to log in again?
Whatman™ Regenerated Cellulose Membranes, Circles, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
10410312 10410219 10410314 10410224 10410380 10410206 10410014 10410314 10410319 10410212 10410014 10410212 10410312 10410206 10410224 10410380 10410214 10410229 10410319 10410214 10410229 10410012 10410012 10410219
10034-992PK 350.55 USD InStock
CA11013-502 10034-992 10034-990 CA11013-246 97010-106 CA11013-520 97010-104 97010-102 CA11013-042 97010-100 10034-998 97010-098 10034-996 97010-096 10034-994 97010-094 CA11012-912 CA97010-094 CA97010-096 CA97010-098 CA97010-100 CA97010-102 CA97010-104 CA97010-106
Whatman™ Regenerated Cellulose Membranes, Circles, Whatman products (Cytiva)
Filters Membrane Filters
Cytiva's Whatman regenerated cellulose membrane filters are microporous cellulose acetate filters made without wetting agents. RC60 offers low extractables and wide compatibility in both organic and aqueous media.
  • Spontaneous wetting with very good wet strength
  • Chemical resistance suitable for a wide range of aqueous and organic media
  • Mechanically stable with low protein binding
  • Low extractable levels to minimize sample contamination
  • Can be sterilized by all common methods
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty