Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glas-Col® Flexible Fabric Heating Mantle, Antylia Scientific

Supplier: Antylia Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Glas-Col®
36225-50 36225-02 36225-60 36226-10 36225-70 36226-30 36225-06 36225-40 36225-75 36225-04 36225-00 36226-20 36226-25 36225-30 36225-10
97010-924EA 539.92 USD InStock
97010-924 97010-956 97010-988 97010-976 97010-932 97010-920 97010-964 97010-984 97010-940 97010-972 97010-992 97010-980 97010-928 97010-916 97010-948
Glas-Col® Flexible Fabric Heating Mantle, Antylia Scientific
Heaters Heating Mantles
Heating Mantles feature insulated glass fabric that conforms to unusual vessel shapes.

  • Maximum temperature of 842°F (450°C)
  • Require a controlled electrical power source to prevent overheatin
  • Models for 120 VAC operation are CSA-certified

Hemispherical Heating Mantles feature insulated glass fabric that conforms to unusual vessel shapes. Steel supports (sold separately) are required to support mantle and allow attachment to ring stands. Supports have fabric strap basket to hold mantles;do not place mantle directly on ring stand.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty