Would you like to log in again?
pH and ATC Pens, Large Display, Sper Scientific
Supplier: Sper Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
850050 850051 850052
97011-762EA 210.71 USD InStock
97011-762 97011-764 97011-760
pH and ATC Pens, Large Display, Sper Scientific
Electrochemical Meters pH and Multi-Parameter Meters
Easy to carry, simple to operate and sufficient for most field pH tests.
  • Highly stable and accurate readings with a large LCD display
  • Designed for simple one-hand operation
  • Features hold function, and auto power off with low battery indicator
Automatically calibrates to pH 4, 7, 10 buffers and can be adjusted to recognize non-standard buffers as well. The case is IP65 waterproof, and will float if accidentally dropped into the water.
Large Display pH Pen: Has a Range of 2 to 12 with an Accuracy of ±0.3 pH and is the perfect instrument for economical spot sampling of pH.
Large Display ATC pH Pen: Has a Range of 0 to 14 with an Accuracy of ±0.2 pH and is the right instrument for spot sampling of pH and temperature. Displays temperature in both °C and °F.
Large Display ATC 0.01 pH Pen: Has a Range of 0 to 14 with an Accuracy of ±0.05 pH and is the ideal instrument when you need laboratory accuracy in the field for sampling of pH and temperature. Displays temperature in both °C and °F.
Ordering information: Comes with 4 button-cell batteries and a built-in electrode kept moist with a protective cap. 1 year warranty.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty