Would you like to log in again?
Data Acquisition Software, Sper Scientific
Supplier: Sper Scientific
840052
97011-798EA 80.54 USD InStock
97011-798
Data Acquisition Software, Sper Scientific
Software Laboratory Software
Software downloads data to your PC.
  • Provides further analysis by receiving the data stream from your meter
  • Data is saved in a text format (.txt) and can easily imported into spreadsheets, databases or word processing programs
Up to 16000 sample readings with real-time clock memory. Programmable sample interval from 1 second to 3,600 seconds. Unit displayed with changing setting. User-defined maximum (High) and minimum (Low) alarm setting. Includes one CD Rom and this operation manual.
System Requirements include Win 95 or higher operating system, 486 PC or higher withat least one serial port, RS232 Cable model 840054 sold separately above.
Works with meter models 800046, 840003, 840028, 840080, 840081, 840082, 840083, and 840085.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty