Would you like to log in again?
Sound Level Pens, Sper Scientific
Supplier: Sper Scientific
840018 840018C
97011-814EA 177.27 USD InStock
97011-814 97011-816
Sound Level Pens, Sper Scientific
Sound Meters
Type 2 sound meter with one-button operation.
The Sper Scientific Sound Level Pen features a type 2 internal calibrator, one-button operation, low cost, auto-ranging and max hold in a mini size.
The Sper Scientific Sound Level Pen is the smallest, lightest, easiest to carry sound meter in the world. Despite its small size, the Sound Level Pen is a full function type 2 (Meets ANSI S1.4 Type 2 and IEC61672-1 Class 2) sound meter perfect for testing OSHA requirements and most other sound meter applications. Auto-ranging with simple one-button operation. Press once to begin reading sound levels. Press the button a second time and the display holds the maximum reading, updating as new maximums are reached. Hold the button for 2 seconds to turn the unit off. Measures the 32–130 dB range in the "A" scale, with an accuracy of ±1.5 dB. The display provides 0.1 dB resolution, under range, over range and low battery indicators. The Sound Level Pen calibrates electronically to its own internal oscillator (94dB at 1kHz) or external 2 Pt. Acoustical Calibrator 850016.
Certifications: N.I.S.T. traceable certificate of calibration (for certified model only).
Ordering information: Comes ready to use with two AAA batteries, instructions, calibration tool, and a wind screen. Five year warranty.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty