Would you like to log in again?
Clinical Refractometer, Sper Scientific
Supplier: Sper Scientific
300005
97011-834EA 297 USD InStock
97011-834
Clinical Refractometer, Sper Scientific
Refractometers Handheld Refractometers
The Sper Scientific Clinical Refractometer measures urine specific gravity, or provides the refractive index in seconds
To operate, simply place a drop or two of sample on the prism and read the results
Ordering information: The Clinical Refractometer is supplied with a hard carrying case, calibration tool, and 5-year warranty.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty