Would you like to log in again?
Calibration Reference Materials for Autosamplers, DSC, PerkinElmer
Supplier: Perkin Elmer
03190033 03190034 03190033
97012-532EA 230.83 USD InStock
CA97012-532 97012-532 97012-270
Calibration Reference Materials for Autosamplers, DSC, PerkinElmer
Standards Alloys and Metal Ore Standards
These calibration reference materials are 99.999% pure
DSC requires elements of this purity so that the instrument can be accurately calibrated.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty