Would you like to log in again?
High Pressure Sample Pans, PerkinElmer
Supplier: Perkin Elmer
B0182901 B0182901 B0182902 03190029 B0182902 03190218 03190029
97013-784EA 810.3 USD InStock
97013-784 97012-954 CA97013-784 CA97013-690 CA97012-954 97012-540 97013-690
High Pressure Sample Pans, PerkinElmer
Trays/Dishes/Pots Laboratory Trays/Dishes
Pans contain the sample in a sealed environment throughout an experiment.
  • Pressure ranges from 24 to 150 bar
The sealing suppresses the vaporization of a solvent or contains a volatile reaction product, thereby eliminating the interfering effects of the heat of vaporization.
The stainless steel pans with covers and seals are reusable and includes 5 pans and 20 gold-plated seals.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty