Would you like to log in again?
Sample Capillary Adapter for GemCone and GemTip Nebulizers, PerkinElmer
Supplier: PerkinElmer
N0371505 N0371505
97012-744EA 38.06 USD InStock
CA97012-744 97012-744
Sample Capillary Adapter for GemCone and GemTip Nebulizers, PerkinElmer
Chromatography Supplies Chromatography Spare Parts
This Capillary Adapter is designed for use with AAnalyst® 100/300/200/400/700/800 AA series instruments, as well as all Avio and Optima™ ICP-OES series instruments.
This Capillary Adapter, or Gland Nut, is included in the GemTip High-Sensitivity Plastic Body Nebulizer Complete Assembly for AAnalyst® and the Universal GemTip Corrosion-Resistant Plastic Body Nebulizer Complete Assembly for AAnalyst®. It is used in the GemCone Low Flow Nebulizer and the GemCone High Dissolved Solids Nebulizer.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty