Would you like to log in again?
Demountable Quartz Torch, 1-Slot for Optima 3x00 DV, PerkinElmer
Supplier: PerkinElmer
N0691662 N0691662
97012-918EA 814.13 USD InStock
CA97012-918 97012-918
Demountable Quartz Torch, 1-Slot for Optima 3x00 DV, PerkinElmer
Chromatography Supplies Chromatography Spare Parts
This 1-slot quartz torch is designed for use with Optima™ 3x00 DV ICP-OES series instruments.
The demountable 1-slot torch design uses one-piece quartz tubing for plasma and auxiliary gas flow, and is the standard torch for aqueous solutions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty