Would you like to log in again?
Jet Replacement Tool for Flame Ionization Detector (FID), Auto-Ignite, PerkinElmer
Supplier: Perkin Elmer
N6101297
97013-060EA 158.99 USD InStock
97013-060
Jet Replacement Tool for Flame Ionization Detector (FID), Auto-Ignite, PerkinElmer
Chromatography Supplies Chromatography Spare Parts
This jet replacement nut driver uses to removing jet from auto ignite FID body.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty