Would you like to log in again?
Lumina™ Barium (Ba) Hollow Cathode Lamp, PerkinElmer
Supplier: PERKINELMER U.S. LLC
Lumina™
N3050109 N3050109
97013-278EA 1807.77 USD InStock
97013-278 CA97013-278
Lumina™ Barium (Ba) Hollow Cathode Lamp, PerkinElmer
Lights Hollow Cathode Lamps
Series N30501XX Lumina™ 2" (50 mm) diameter single-element lamps are designed to be used with the PinAAcle™ and AAnalyst™ Atomic Absorption spectrometer series of instruments.
  • This element requires the use of a nitrous oxide-acetylene flame
  • Lumina™ hollow cathode lamps may also be used with all earlier AA spectrometers that do not have automatic code reading capability by using an appropriate adapter cable
Single Element Lumina™ Hollow Cathode Lamp (HCL) for the detection of elemental Barium (Ba). Every genuine PerkinElmer lamp is designed for use with and tested on spectrometers to assure compatibility and the highest performance. A wide range of single-element and multi-element HCLs are available, and are ideal for determining most elements by atomic absorption spectroscopy. To prolong the life of a Lumina™ Hollow Cathode Lamp (HCL), lamps are manufactured with a larger internal volume so that a greater supply of fill gas at optimum pressure is available. The larger the lamp, the greater the inert gas volume, and therefore the longer the lamp lifetime.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty