Would you like to log in again?
Screw Top Vials, 10 mm, PerkinElmer
Supplier: PerkinElmer
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
N9306057 N9306053 N9306057 N9306053
97013-862PK 67.61 USD InStock
97013-862 CA97013-862 97013-858 CA97013-858
Screw Top Vials, 10 mm, PerkinElmer
Vials
Vials are manufactured from Type I borosilicate glass, which meets all USP, JP, and EP Pharmacopeia requirements.
  • Clear or amber color
  • Resistant to high temperature
  • Chemical resistant to acidic, neutral, and alkali solutions
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty