Would you like to log in again?
Orion™ Green pH Combination Electrodes, Thermo Scientific
Supplier: Thermo Scientific
GD9156BNWP GD9106BNWP GS9106BNWP
97014-788EA 369.55 USD InStock
97014-788 97014-784 97014-782
Orion™ Green pH Combination Electrodes, Thermo Scientific
Electrodes pH Electrodes
The Green electrodes are the first laboratory glass-bulb pH electrodes to meet all the requirements of RoHS.
  • Choice of single-junction models for clean-water measurements, or double-junction models for dirty-water testing
  • Choice of refillable or low-maintenance gel-filled models
  • No disposal issues or disposal cost
  • pH range is 0 to 14
  • Durable epoxy body
These environmentally friendly pH combination electrodes with epoxy body and waterproof BNC connector contain no lead, mercury or other hazardous substance.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty