Would you like to log in again?
Chromolith® CapRod® Capillary Columns, MilliporeSigma
Supplier: MilliporeSigma
Chromolith® CapRod®
1.52031.0001 1.50424.0001 1.50405.0001 1.50400.0001 1.50402.0001 1.50407.0001 1.50409.0001 1.50426.0001 1.50402.0001 1.50426.0001 1.50404.0001 1.50424.0001 1.50405.0001 1.50402.0001 1.50404.0001 1.50403.0001 1.50407.0001 1.50409.0001 1.50400.0001 1.50403.0001 1.52031.0001
10144-110EA 2153.77 USD InStock
CA1.50402.0001 CA97015-678 10144-110 97015-678 CA1.50426.0001 10144-112 EM1.52031.0001 CA1.50424.0001 10143-570 48219-914 97015-680 EM1.50402.0001 CA1.52031.0001 CA1.50404.0001 CA97015-674 97015-674 CA97015-684 97015-684 CA97015-682 CA97015-680 97015-682
Chromolith® CapRod® Capillary Columns, MilliporeSigma
Chromatography Columns
All Chromolith columns are made of monolithic silica with a characteristic bimodal pore structure.
  • Typical flow rates are above 1 µl/min
Chromolith CapRod is a capillary column that combines the speed of monolithic silica technology with the sensitivity of nano-LC, hence enabling new productivity levels for high-throughput proteomics LC applications. In contrast to microparticulate columns Chromolith CapRod can be operated at significantly higher flow rates without loss of performance. Separations can be achieved at 1 to 3 µl/min compared to 200 to 400 nl/min for microparticulate columns.
The monolithic Trapping Column is a capillary trap that is used to concentrate samples before transfering them to the analytical Chromolith CapRod column.
Chromolith 3 mm columns have macropores with 2 µm diameter, giving a column efficiency exceeding 80000 plates/meter. The mesopores are 13 nm (130 Å), the surface modification is octadecylsilan with full end-capping.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty