Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Orion™ RDO™ Dissolved Oxygen Probe, Thermo Scientific
Supplier: Thermo Scientific
Orion™
087030MD 087100MD 087010MD 087050MD 087020MD
97015-858EA 1729.91 USD InStock
97015-858 97015-856 97015-864 97015-862 97015-860
Orion™ RDO™ Dissolved Oxygen Probe, Thermo Scientific
Electrodes Dissolved Oxygen Electrodes
Measure dissolved oxygen levels without the difficulties of membrane replacement by using the Orion™ RDO™ Dissolved Oxygen Probe.
  • Utilizes the latest lumiphore technology for measuring DO
  • Once-a-year maintenance with a simple cap replacement
  • Sensor cable lengths of 3, 6, 10, 15, or 30 m (10, 20, 30, 50, or 100')
  • IP68-rated, waterproof
The Orion™ RDO™ dissolved oxygen probe offers a dissolved oxygen concentration range of 0.00 to 20.0mg/l, and a dissolved oxygen saturation range of 0.0 to 200%. Each probe order includes RDO probe, optical cap, stainless steel sensor guard, and calibration sleeve.
RDO (Rugged Dissolved Oxygen) Probe uses optical DO technology for problem-free measurement.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty