Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
iLog® Temperature Loggers, Cryopak Verification Technologies
Supplier: Cryopak Verification Technologies
EI-2E-C-32-L EI-2E-DC-32-L EI-IN-D-32-L EI-2E-N-32-L EI-2E-DZ-32-L EI-1E-D-32-L EI-2E-DN-32-L EI-2E-NZ-32-L EI-2E-D-32-L EI-2E-CZ-32-L EI-2E-Z-32-L EI-1E-DC-32-L EI-2E-CN-32-L
97017-216EA 1207.68 USD InStock
97017-216 97017-238 97017-204 10033-860 10033-858 97017-198 10033-856 10033-866 10033-854 10033-864 10033-852 97017-200 10033-862
iLog® Temperature Loggers, Cryopak Verification Technologies
Data Loggers
The user programmable iLog data logger provides technological abilities for monitoring temperature sensitive products while supporting compliance requirements. The case is made from Polycarbonate.
  • Large LCD display with multi-functional viewing
  • Visual and audible alarms
  • Bookmark capabilities
  • Wide temperature range (high heat, ambient, refrigerated, frozen and dry ice)
  • High alarm; low alarm configuration
  • Two year warranty
Detachable, customizable external sensor lengths and probe styles.
Certifications: ISO9001:2008, CE, RoHS, EN 12830.
Ordering information: A temperature/humidity logger is available; contact your Avantor representative for more information.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty