Would you like to log in again?
VWR® Three-Sided Enclosure Carts with Round Posts
Supplier: VWR
VWR®
R243672CTS4V R186072CTS4V R183663CTS4V R244863CTS4V R243663CTS4V R246072CTS4V R246063CTS4V
97017-634EA 2564.78 USD InStock
97017-634 97017-610 97017-628 97017-636 97017-626 97017-630 97017-620
VWR® Three-Sided Enclosure Carts with Round Posts
Carts
Detachable wire mesh panels allow secure handling of objects during transport.
  • Secure handling of objects
  • Truss-reinforced edges
  • Exceptional durability
  • Adjustable shelf height
  • Round post design
Wire shelves and components feature high-quality, bright chrome plating. Shelves have truss-reinforced edges for exceptional durability. Posts also feature groove rings in 2.5 cm (1") increments for shelf height adjustability. Carts can support up to 544kg (1200lbs.) when weight is evenly distributed.
Ordering information: Supplied with four shelves, four corner posts, four 12.7 cm (5") swivel casters, and detachable panels.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty