Would you like to log in again?
BULK SPE FLORISIL 100G
Supplier: Agela Technologies
Cleanert Florisil 100G40-60 U80-100 U^ Pore Size: 80A; Surface Area: 291M2/G.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BULK SPE FLORISIL 100G 97018-016 FS0100 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty