Would you like to log in again?
COL SPE PS 200MG 6ML PK30
Supplier: Agela Technologies
Cleanert Ps 200Mg 6mL 40 U^ Pore Size: 70A; Volume Of Pore: 1.2Cm2/G; Surface Area: 600M2/G.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COL SPE PS 200MG 6ML PK30 97018-176 PS2006 Pack of 30 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty