Would you like to log in again?
VWR® Talon® PVC Coated Open Extension Rings
Supplier: VWR
VWR® Talon®
80863386 80863385 80863384
97019-056EA 106.66 USD InStock
97019-056 97019-054 97019-058
VWR® Talon® PVC Coated Open Extension Rings
Support Rings
Ideal for supporting funnels, round-bottom flasks, reaction vessels, and other apparatuses requiring lower support.
  • PVC coating
  • Fully adjustable depth
  • Capable of supporting a wide range of glassware
These PVC-coated aluminum rings feature an opening that allows simple removal of sample containers. The long extension arm permits depth adjustment of the open ring from the lab frame or ring stand. PVC coating protects glassware.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty