Would you like to log in again?
Deck-Mounted Gooseneck Mixing Faucets, with Nylon Hooded Handles, WaterSaver Faucet
Supplier: WaterSaver Faucet
CT414-8VB CT414VB CT4414 CT4414-8VB CT4414VB CT414 CT414-8
97021-296EA 397.3 USD InStock
97021-296 59382-092 97021-298 59382-094 VFT-0414-00 VFT-0414-VB 97021-292
Deck-Mounted Gooseneck Mixing Faucets, with Nylon Hooded Handles, WaterSaver Faucet
Faucets and Sinks Faucets
Deck-mounted mixing faucets are made of white epoxy/polyester hybrid powder-coated forged brass. They have a serrated hose end, black or color-coded nylon hooded handles, and 1cm (3/8") copper tubes on inlets. Faucets are available with 15.2cm (6") or 20.3cm (8") spread swinging gooseneck, with or without a vacuum breaker. Faucet valves are self-contained renewable units with replaceable stainless steel seats.
  • Faucet valves are self-contained renewable units with replaceable stainless steel seats
They have a serrated hose end, black or color-coded nylon hooded handles, and 1 cm (3/8") copper tubes on inlets. Faucets are available with 15.2 cm (6") or 20.3 cm (8") spread swinging gooseneck, with or without a vacuum breaker.
Certifications: CSA certified. Comply with ANSI/ASME A112.18.1M.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty