Would you like to log in again?
Rotor for Centrifuges 5418 R
Supplier: Eppendorf
022652061
97021-820EA 1218.77 USD InStock
97021-820
Rotor for Centrifuges 5418 R
Centrifuges Rotors
Eppendorf rotor for centrifuges 5418 R, precision redefined. Tailor your centrifugation with varied capacities and speeds. Accessorise with adaptors and buckets for versatility in molecular biology and life science applications.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotor, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 minutes)
  • Accessories (adaptors and buckets) further customize your centrifuge setup
Experience precision in centrifugation with Eppendorf rotor for centrifuges 5418 R. Offering a spectrum of capacities and speeds, these rotors cater to diverse applications from microcentrifugation to larger sample volumes. Accessorise your setup with adaptors and buckets, enhancing versatility for specific molecular biology and life science workflows. Achieve optimal sample separation, ensuring reproducibility in your experiments. Explore the array of options to customize your centrifuge 5418 R for reliable and efficient performance.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty