Would you like to log in again?
ASTM Class 1 Weight Sets, METTLER TOLEDO®
Supplier: Mettler-Toledo
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
11124018 11124001 11124015 11124014 11124003 11124007 11124009 11124005 11124018 11124002 11124007 11124004 11124004 11124000 11124008 11124006 11124019 11124014 11124019 11124005 11124006 11124008 11124009 11124001 11124015 11124002 11124003
97021-838EA 905 USD InStock
CA97021-848 97021-838 CA97021-846 97021-860 CA97021-852 97021-842 97021-864 CA97021-850 97021-862 97021-840 CA97021-856 97021-846 97021-844 CA97021-854 97021-848 CA97021-858 97021-850 97021-854 CA97021-840 CA97021-862 CA97021-860 97021-852 97021-858 CA97021-844 97021-856 CA97021-842 CA97021-864
ASTM Class 1 Weight Sets, METTLER TOLEDO®
Weights Calibration Weights
These ASTM weight sets with adjusting cavity are made of vaccum-melted stainless steel, with an electropolished finish.
  • Electrolytic polishing ensures glossy surfaces for anti-adhesion effects
  • Exclusive material has a density of 7.9 kg/dm3
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty