Would you like to log in again?
FILTER TEXAS RED/CY3.5 SET
Supplier: VWR International
Fluorescence Filter Texas Red/Cy3.5 Set^ Exciter D560/40X, Dichroic 595Dclp, Emitter D630/60M.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
FILTER TEXAS RED/CY3.5 SET 97021-910 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty