Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

AE31E LED Inverted Compound Microscopes, Motic Instruments

Supplier: Motic
Immobiline™
1101000203431 1101000203291 1101000200111 1101000203301 1100100200574 1100100200562 1100100200574 1101000202651 1101000203251
97021-928EA 1149.81 USD InStock
97021-928 CA97021-924 CA97021-908 10767-574 CA10769-640 CA97021-906 10025-352 10767-576 CA97021-910
AE31E LED Inverted Compound Microscopes, Motic Instruments
Microscopes
With a wide range of potential applications, Motic’s AE31 Elite microscope is at home in both research and routine environments.

  • Siedentopf binocular tube 45° inclined (binocular model)
  • Siedentopf trinocular tube 45° inclined [light split: 20/80] (trinocular model)
  • WF PL 10×/22 mm high eyepoint eyepiece, with diopter adjustment and rubber eyecup
  • Side facing quintuple nosepiece
  • CCIS Plan Achromat objective PL4×
  • CCIS Plan Achromat phase objective PL Ph10×, LWD PL Ph20×
  • ELWD N.A. 0.30 (W.D. 72 mm) condenser
  • Phase slider Ph0, BF, Ph1
  • Phase centering telescope
  • Plain stage, glass and metal stage inserts
  • 3W LED illumination with intensity control
  • 45 mm blue, ground glass and green interference filter
  • Power cord, allen hexagonal key, vinyl dust cover
  • Universal power supply 100 - 240 V UL
  • Modular system (trinocular model)

Combining the AE31 Elite with a Moticam digital microscopy camera provides stunning publication quality images and transforms the AE31 into a complete documentation and analysis workstation. The extra-long working distance of 72mm easily accommodates most Petri dishes and culture flasks. This distance is easily increased to 184mm by removing the lower part of the condenser. The available epi-fluoresence attachment is compatible with a wide range of filters, including DAPI, FITC, GFP, and Cy3, among others. The AE31 Elite provides a significant improvement in optical performance and versatility.

The extended focus on the practical aspects allows the AE31 Elite to provide excellent optical quality and unmatched operational convenience when compared to similarly priced units. The included plan achromatic objectives and built-in Koehler illumination provide crisp, evenly illuminated images with high image contrast and perfect field flatness up to F.N.22. Available upgrade options such as epi-fluorescence attachments and LED lighting modules ensure that the AE31 Elite will meet requirements and grow with the lab for years to come.

Ordering information: Microscope is supplied with objectives, stage accessories, Epi-fluorescence attachments. Microscope is customizable with various accessories. For all available options, please contact your local Avantor sales representative. All Motic products come with a one-year electrical and five-year mechanical warranty.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty