Would you like to log in again?
COLUMN PREP AQ C18 20X150
Supplier: Agela Technologies
Preparative HPLC column AQ-C18^ a reverse phase having enhanced retention for polar compounds and compatible with 100 water; Very large sample loading; 380m2/g, C%=18; 100A, 10um.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN PREP AQ C18 20X150 97022-044 VA901520-0 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty