Would you like to log in again?
STANDARD CELL HOLDER
Supplier: PERKINELMER U.S. LLC
Standard Cell Holder^ Accepts one rectangular or microcell with 10 mm pathlength (as standard holder for Lambda instruments included in shipment).
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
STANDARD CELL HOLDER 97024-462 B0505071 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty