Would you like to log in again?
Conical Bottom Autosampler Tubes with Graduation, PerkinElmer
Supplier: Perkin Elmer
B0193233 B0193233
97024-502PK 362.98 USD InStock
97024-502 CA97024-502
Conical Bottom Autosampler Tubes with Graduation, PerkinElmer
Tubes Test Tubes
These standard translucent polypropylene tubes are designed to provide excellent chemical resistance ensuring an accurate analysis.
  • Compatible with PerkinElmer Autosamplers for AA, ICP-OES and ICP-MS instruments
  • Tubes are graduated for ease-of-use
  • Writing patch area for sample identification
These are the standard autosampler tubes for the PerkinElmer S10 autosampler.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty