Would you like to log in again?
Fixed Angle Reflectance Accessory, PerkinElmer
Supplier: PerkinElmer
B0086703 B0086703
97024-528EA 10683.01 USD InStock
CA97024-528 97024-528
Fixed Angle Reflectance Accessory, PerkinElmer
Chromatography Supplies Chromatography Spare Parts
For reflectance or thickness measurements. Use with LAMBDA 25/35/45/650/750/850/950/1050.
  • 6° angle of light incidence
  • Accommodates samples from 20×20 mm to 100×100 mm
  • Wavelength range is 175 to 3330 nm
The fixed angle reflectance accessory locates in the standard sample compartment for routine relative reflectance measurements at a 6° fixed angle of incidence.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty