Would you like to log in again?
Value ATR Top Plates, PerkinElmer
Supplier: Perkin Elmer
PerkinElmer®
L1360267 L1360273 L1360271 L1360266 L1360269 L1360269 L1360271 L1360268 L1360270 L1360270 L1360272 L1360273 L1360268 L1360272 L1360267 L1360266
97024-608EA 1826.37 USD InStock
CA97024-606 97024-608 CA97024-608 97024-614 CA97024-602 97024-604 CA97024-612 CA97024-604 CA97024-614 97024-606 97024-610 97024-600 97024-612 CA97024-600 97024-602 CA97024-610
Value ATR Top Plates, PerkinElmer
Trays/Dishes/Pots Laboratory Trays/Dishes
Value HATR trough/flat top-plates allow for simple sampling of liquids, solids, polymer films, powders, pastes, and gels.
Zinc Selenide crystal is designed for general purpose HATR sampling.
Caution: Avoid use with complexing agents, strong acids, bases, and oxidizers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty