Would you like to log in again?
KBr Disc Starter Pack for Solids, Spectrum RX1 System, PerkinElmer
Supplier: PerkinElmer
L1272266 L1272265
97024-650EA 12294.22 USD InStock
97024-650 97024-652
KBr Disc Starter Pack for Solids, Spectrum RX1 System, PerkinElmer
Presses
The complete pack contains an evacuable potassium bromide die for the preparation of 13mm diameter pellets of the highest quality and clarity, a pellet holder and adjustable mount for precise mounting of samples in your Spectrum RX, the easy-to-operate 15-tonne hydraulic press and 100g of research-grade KBr powder.
Ordering information: Starter pack is available with or without the hydraulic press.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty