Would you like to log in again?
Caps for TrayCell™, PerkinElmer
Supplier: Perkin Elmer
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
L7110292 L7110293 L7110290 L7110291 L7110293 L7110291 L7110290 L7110292
97024-720EA 1314.8 USD InStock
97024-720 CA97024-720 CA97024-716 97024-718 CA97024-718 97024-714 CA97024-714 97024-716
Caps for TrayCell™, PerkinElmer
Closures Test/Sample Tube Closures
The TrayCell™ caps are fiber-optic ultra-micro measuring cells designed for the UV/Vis analysis of DNA/RNA and proteins.
  • Suitable for almost all spectrophotometers
  • Ideally suited for very small measurement volumes: 0.7 to 10 L
  • Trouble-free measurement of the sample at different optical path lengths
  • Fast and simple cleaning of the optics before measuring the next sample
  • Excellent reproducibility and samples can be reused after the measurement simply by piping them off
The dimensions of the TrayCell™ are equivalent to a standard cuvette in order to work in most spectrophotometers. Caps with dilution factors: 5, 10, 50 and 100.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty