Would you like to log in again?
FINE POINT GRIP M6 THREAD 8MM
Supplier: SEALS USA INC
8mm Wide Fine Point Grip^ M6 Thread for FGE-X / FGV-XY force gauges (2-200 lbs). Used for small parts Pull testing
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FINE POINT GRIP M6 THREAD 8MM 97024-832 FG-M6FPG8 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty