Would you like to log in again?
JACOB S CHUCK GRIP
Supplier: SEALS USA INC
Jacobs Chuck Style Grip^ M6 Thread, for FGE-X / FGV-XY force gauges (2-100 lbs) Used for gripping round or thin, 0.2in up to 0.20in dia parts.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
JACOB S CHUCK GRIP 97024-838 FG-M6JAC5U Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty