Would you like to log in again?
COMPRESSION PLATE
Supplier: SEALS USA INC
100mm diameter Compression plate^ 6M thread used for compression testing of bottles, jars, packages. Black metal finish. Threads to load sensor of FGE-X, FGV-XY force gauges
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COMPRESSION PLATE 97024-852 FG-M6COMP100U Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty