Would you like to log in again?
TENSILE GRIP
Supplier: SEALS USA INC
Heavy-Duty Tensile Grip^ M10 Thread for FGE-200HX, FGE/V-500HX force gauge, High capacity,tightened jaw faces can accept samples 1.25in (31.75mm) wide and 0.25in (6.35mm) thick. Contact faces are serrated hardened tool steel
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TENSILE GRIP 97024-890 FG-M10TEN7U Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty