Would you like to log in again?
CLEVIS GRIP
Supplier: SEALS USA INC
Clevis Grip^ w/base Odd shapped parts, tension springs, Concave shape of pins assures centering of parts in the grips under tension loads. Throat clearance w/ pins is 0.5in, can accept samples up to 0.375in wide. for test stand mounting
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CLEVIS GRIP 97024-934 FG-CLV10B Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty