Would you like to log in again?
SET OF 30-MM FIXTURE PEGS 4PC
Supplier: SEALS USA INC
30mm High Fixture Pegs^ 4pcs, Replacement pegs for TNP torque meter. Pegs are removeable, adjustable for various size samples: bottles, jars, motors, tools, solenoids, gears
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SET OF 30-MM FIXTURE PEGS 4PC 97024-984 TNP-TP30 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty