Would you like to log in again?
VWR® Personal-Sized Incubator
Supplier: VWR
VWR®
I5110-VWR-230V I5110-VWR
97025-630EA 675.38 USD InStock
97025-630 97025-632
VWR® Personal-Sized Incubator
Incubators Compact Benchtop Incubators
These personal-sized incubators are ideal for use in microbiology or hematology.
  • Compact Size
  • Economical
  • Corrosion-Resistant Interior Chamber
  • Nontoxic, Optional Thermometer
  • Metal Construction
The housing and door frame are entirely metal
The top of the unit features a grommet hole that accommodates a thermometer for an accurate temperature setting. A port in the rear of the unit allows for an electrical cord to be routed through the back.
A Plexiglas window in the door provides full visibility into the corrosion-resistant interior chamber. The included shelf can be adjusted to three different levels. Heating elements are incorporated into the housing bottom and sides to ensure temperature stability and uniformity.
Accessories information: Additional shelves are available separately to increase storage capacity. The optional thermometer is non-toxic and coated in fluoropolymer resin. It offers a resolution of 1°C and is 30.5 cm (12") long with a 7.6 cm (3") immersion.
Ordering information: The incubator comes with a two-year limited warranty.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty