Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Presence/Absence Test, Hach
Supplier: Hach
2323250 2323250
97026-308PK 581.78 USD InStock
97026-308 CA97026-308
Presence/Absence Test, Hach
Microbiology Test Systems Microorganism Tests
Presence/Absence test for coliforms; Disposable.
Save time and simplify lab and field testing with Hach prepared P/A media
Ordering information: 50/pk.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty