Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
BART Test for Acid Producing Bacteria, Hach
Supplier: Hach
2831409 2831409
97026-408PK 370.72 USD InStock
97026-408 CA97026-408
BART Test for Acid Producing Bacteria, Hach
Microbiology Test Systems Microorganism Tests
BART biodetectors are excellent diagnostic tools to help identify the presence and activity of various bacteria.
Simple yet effective method for monitoring the population size and/or activity of specific groups of bacteria.
Easy to use, requiring no elaborate or costly equipment and no specialized training. Effective and affordable tests are easy to interpret and can be performed at room temperature in virtually any environment.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty