Would you like to log in again?
Phosphate Standard Solution, 1 mg/L as PO₄ (NIST), 500 ml, Hach
Supplier: Hach
256949 256949
97026-656EA 76.68 USD InStock
97026-656 CA97026-656
Phosphate Standard Solution, 1 mg/L as PO₄ (NIST), 500 ml, Hach
Standards Environmental Analysis Standards
For accuracy checks in phosphate (phosphorus) determinations.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty