Would you like to log in again?
Digital Titrator, Hach
Supplier: Hach
1690001 1690001
97026-774EA 510.97 USD InStock
97026-774 CA97026-774
Digital Titrator, Hach
Titrators
Digital Titrator, 0.00125 ml/digit, with case, manual, and five straight delivery tubes.
  • Accurate results
  • when and where you need them
  • Fast and flexible
  • Versatile
  • Economical
  • EndPoint easier to determine
Accessories information: Titration delivery tubes for use with Hach digital titrator with 90 degree/right angle bend with J-hook tip.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty