Would you like to log in again?
Free Chlorine Test Strips, 0-600 mg/L, Hach
Supplier: Hach
2890200 2890200
97026-782EA 69.38 USD InStock
CA2890200 97026-782
Free Chlorine Test Strips, 0-600 mg/L, Hach
Environmental Test Kits
These test strips are manufactured with a unique reagent pad at the tip, using proven chemistries based on standard reference methods.
  • Ideal for anyone performing semiquantitative prescreenings, spotchecks, or field tests
  • Simplify testing and get results in less than 1 minute, in the field or lab
  • Eliminate chemical handling and cleanup
  • Reduce guesswork by screening samples
Just dip the strip, wait until color development occurs, and compare the color of the reacted pad to the chart printed on the bottle
*mg/L unless otherwise noted; ppb = μg/L; ppm = mg/L; gpg = grains per gallon; 1 gpg = 17.1 mg/L or 17.1 ppm.
*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty