Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Corning® Spin-X® UF Concentrators, Corning

Supplier: Corning

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Corning®
431491 431486 431477 431487 431489 431490 431484 431482 431483 431478 431488 431480 431481 431485 431479
97027-896CS 159.05 USD InStock
97027-896 97027-894 97027-892 97027-890 97027-904 97027-914 97027-902 97027-912 97027-900 97027-888 97027-910 97027-898 97027-908 97027-906 97027-916
Corning® Spin-X® UF Concentrators, Corning
Filters Centrifugal Filters
These non sterile centrifugal concentrators offer a simple, one step procedure for concentrating or desalting proteins and other biomolecules with 90% or better recovery.

  • Vertical membrane design and thin channel filtration chamber minimises membrane fouling and provides fast, high speed concentrating, even with particle laden solutions
  • Integrated dead stop design eliminates risk of spinning to dryness; no respinning necessary
  • Low binding polyethersulfone (PES) membranes are available with five molecular weight cut-offs (MWCO): 5000, 10000, 30000, 50000 and 100000 to meet all of your concentrating needs
  • Choose an MWCO ½ to ⅓ smaller than the protein to be concentrated
  • The MWCO and graduations are printed right on the side of the concentrator tube to avoid mix-ups

Other applications include removal of labeled amino acids and nucleotides, HPLC sample preparation, deproteinization of samples, recovery of biomolecules from cell culture supernatants and lysates, and concentrating virus from cell culture supernatants.

Choice of three sizes for better flexibility:
- Spin-X UF 500 for samples up to 500 µl
- Spin-X UF 6 for samples up to 6 ml
- Spin-X UF 20 for samples up to 20 ml, 14 ml if using fixed angle rotors

Ordering information: Spin-X UF 6 and 20 concentrators can be used with either swinging bucket or fixed angle rotors.
Spin-X® UF 20 concentrators have a capacity of 14 ml if using fixed angle rotors.
Spin-X UF 500 concentrators require fixed angle rotors. Rotors are not included. Please contact Avantor for further information.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty