These test sieves consist of a solid stainless steel sieve frame of high stability for reliable sieving results. Sieves are mesh-specific and thus sieve fabric is precisely joined into the frame and tautened.
- High degree of corrosion resistance and easy cleaning
- Permanently tight sieve fabric
- Extensive optical inspection
- Clear and precise labeling of the sieves with full traceability based on individualised laser engraving
- Maximum stability and optimum sealing when used in sieve stacks thanks to the o-ring which is placed in the recess designed for this purpose
Applicable for separation, fractioning, particle size determination
Field of application includes agriculture, biology, chemistry/plastics, engineering, electronics, environment, recycling, food, geology, metallurgy, glass, medicine.
Feed materials like powders, bulk materials, suspensions, cement clinker, chemicals, coffee, construction materials, fertilizers, fillers, flours, grains, metals powders, minerals, nuts, plastics, sand, seeds, soils, washing powder etc.
Certifications: ASTM E 11.