Would you like to log in again?
Cogent™ UDC-Cholesterol, MicroSolv Technology Corporation
Supplier: MicroSolv
Cogent™ UDC-Cholesterol
69069-HG4 69069-HG5 69069-MP150 69069-HG2 69069-15P-1 69269-15P-2 69269-75P-2 69069-P21-150 69069-MP50 69269-10P-2 69269-02P-2 69069-SP100 69269-05P-1 69069-SP150 69069-10P-1 69069-P21-50 69069-HG6 69069-P21-100 69069-7.5P 69069-SP50 69069-P21-250 69069-SP250 69069-05P-2 69069-10P-2 69269-05P-2 69069-15P-2 69069-15P 69069-HG3 69269-05P-3 69269-03P-3 69269-03P-2 69069-MP100 6906925P 69069-10P 69069-10P-3 69069-HG1
75801-198EA 2501.2 USD InStock
75801-198 75801-196 10803-792 75801-194 10803-790 75801-192 75801-190 97055-198 89234-700 10022-594 10022-596 89234-686 97035-002 97035-000 89234-682 89234-684 10022-598 75801-188 75801-186 75801-184 75801-182 75801-180 89220-042 10022-564 97035-010 97034-992 89234-694 97035-130 10803-794 97034-994 10022-566 97034-996 89234-690 10022-568 97034-998 75801-178
Cogent™ UDC-Cholesterol, MicroSolv Technology Corporation
Chromatography Columns
The many advanced features of the Cogent UDC-Cholesterol column produces a very stable, efficient and unique HPLC column with unique selectivity for steroids and other compounds
  • Not endcapped liquid crystal
  • Shape recognition
  • 'Steroids' column
  • TYPE-C™ silica
The Cogent UDC-Cholesterol™, an eleven carbon link with cholesterol attached is bonded to the silica hydride surface with a direct silicon-carbon bond. This phase is more sensitive to heat and solvent composition than most phases due to the liquid crystal nature of the cholesterol moiety. These parameters can be used in shape recognition and can make for very unique selectivity on this very stable and robust column.
Operates in reversed phase (RP), normal phase (NP) and/or aqueous normal phase (ANP), solving many difficult separation problems.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty